Air admittance valve
183-1201
- Description:Air admittance valve, small flange KF DN 10
- Pk:1
These valves are compact air admittance valves with small flange. The VB valve is manually operated. On rotating the knurled screw cap the valve disc is lifted off the seating and air enters through the holes. The valve disc is rotatable. Therefore the valve seat seal is prevented against wear during opening and closing.
Leakage rate: 1×10–6 mbar l/s.