Water purification systems, Select Analyst
Supplier: Purite
Certificates
About this item
The Select Analyst is a compact unit ideal for Laboratory use, delivering a steady supply of > 1MΩ.cm pure water for Laboratory tasks using carbon pre-treatment, reverse osmosis and deionisation.
- RO removes > 98% minerals and > 99% bacteria
- RO boost pump fitted as standard
- LCD colour touch screen panel, visual and audible alarms
- Available with or without integral 20 litre storage
- Semi-automatic clean cycle
- USB port to download event data and upload software updates
- WiFi connection
Typical applications include glassware rinsing, buffers and stains, reagent make-up, and media preparation.
Installation kit and all consumables included for first year's operation.
Delivery: Supplied with alphanumeric display panel, pretreatment cartridge, reverse osmosis module, deionisation module and single tank bib-tap. External tanks required for GP models are also available, please contact your local Avantor office for details.
Specifications
- Catalog No:
- 171-0187
- 171-0282
- 171-0189
- Model:
- Analyst 160
- Analyst 320
- Analyst 320
- Feed water:
- Potable (tap)
- Potable (tap)
- Potable (tap)
- Production flow rate:
- 14,4 - 24 L/h
- 30 - 48 L/h
- 30 - 48 L/h
- TOC content:
- <50 ppb
- <50 ppb
- <50 ppb
- Inorganics @ 25°C:
- >1 MΩ/cm
- >1 MΩ/cm
- >1 MΩ/cm
- Bacteria:
- >99% rejection*
- >99% rejection*
- >99% rejection*
- W×D×H:
- 440×560×750 mm
- 440×560×750 mm
- 440×560×750 mm
- Weight:
- 28 - 43 kg
- 33 - 52 kg
- 33 - 52 kg
Specifications
Cartridges
Product Information
Frequently Bought Together