VWR®, Cheesecloth Wipes
VWRU21910-107
- Packed:200/box
- Colour:White
- L×W:101×101 mm
- Seq:01
- Pk:200
These bleached cloths made from 100% pure cotton of reagent-grade quality and purity are ideal for wiping, cleaning, or drying cuvettes, delicate glassware, syringes, pipettes, and pH electrodes.
- Unique combination of cleanliness, softness, absorbency, and strength
- Will not scratch the most sensitive surfaces
- Absorb six times their weight and absorb both solvents and aqueous solutions
- Inert to even harsh solvents
- Mesh size: grade 10
Wipes 21910-107 can be unfolded to approximately 178×356 mm. Wipes 21910-105 can be unfolded to approximately 457×914 mm.
: Wipes meet cleanliness specifications of USP and Federal Specification CCC-G-101c.
: Supplied in a convenient dispenser box.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904