Order Entry
Ireland
ContactUsLinkComponent
Anti-Wnt5a Polyclonal Antibody
Anti-Wnt5a Polyclonal Antibody
Catalog # BSBTPB9063
Supplier:  Boster Bio
undefined
Anti-Wnt5a Polyclonal Antibody
Catalog # BSBTPB9063
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Wnt5a
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for Wnt-5a/WNT5A detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. Wnt-5a/WNT5A information: Molecular Weight: 42339 MW; Subcellular Localization: Secreted, extracellular space, extracellular matrix; Tissue Specificity: Expression is increased in differentiated thyroid carcinomas compared to normal thyroid tissue and anaplastic thyroid tumors where expression is low or undetectable. Expression is found in thyrocytes but not in stromal cells (at protein level).

Type: Primary
Antigen: Wnt5a
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat