Anti-Ubiquitin Polyclonal Antibody
Catalog # BSBTPB9122
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for Ubiquitin/UBB detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. Ubiquitin/UBB information: Molecular Weight: 25762 MW; Subcellular Localization: Ubiquitin: Cytoplasm . Nucleus .
Type: Primary
Antigen: Ubiquitin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat