Anti-MMP-14 Polyclonal Antibody
BSBTPB9283
- Antibody type:Primary
- Antigen symbol:MMP-14
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Polyclonal antibody for MT1 MMP/MMP14 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. MT1 MMP/MMP14 information: Molecular Weight: 65894 MW; Subcellular Localization: Membrane ; Single-pass type I membrane protein . Melanosome. Cytoplasm. Identified by mass spectrometry in melanosome fractions from stage I to stage IV. Forms a complex with BST2 and localizes to the cytoplasm; Tissue Specificity: Expressed in stromal cells of colon, breast, and head and neck. Expressed in lung tumors.
Type: Primary
Antigen: MMP-14
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human