Anti-Menin Polyclonal Antibody
Catalog # BSBTPB9263
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:Menin
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for Menin/MEN1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. Menin/MEN1 information: Molecular Weight: 68023 MW; Subcellular Localization: Nucleus . Concentrated in nuclear body-like structures. Relocates to the nuclear matrix upon gamma irradiation; Tissue Specificity: Ubiquitous.
Type: Primary
Antigen: Menin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse