Anti-Hsp90 Alpha Polyclonal Antibody
Catalog # BSBTPB9089
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Hsp90 Alpha
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for HSP90/HSP90AA1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. HSP90/HSP90AA1 information: Molecular Weight: 84660 MW; Subcellular Localization: Cytoplasm. Melanosome. Cell membrane. Identified by mass spectrometry in melanosome fractions from stage I to stage IV.
Type: Primary
Antigen: Hsp90 Alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat