Anti-Emmprin Polyclonal Antibody
BSBTPB9247
- Antibody type:Primary
- Antigen name:Emmprin
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Size:100 µg
- Pk:100 µG
Polyclonal antibody for 575 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: Flow Cytometry. Reactive species: Human. 575 information: Molecular Weight: 42200 MW; Subcellular Localization: Cell membrane ; Single-pass type I membrane protein . Melanosome . Colocalizes with SLC16A1 and SLC16A8. Identified by mass spectrometry in melanosome fractions from stage I to stage IV; Tissue Specificity: Present only in vascular endothelium in non- neoplastic regions of the brain, whereas it is present in tumor cells but not in proliferating blood vessels in malignant gliomas.
Type: Primary
Antigen: Emmprin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human