β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form) ≥98%, white powder cell culture reagent
: Nadide, NAD+, Nicotinamide adenine dinucleotide, oxidized
:53-84-9
:
:C₂₁H₂₇N₇O₁₄P₂
:MFCD00036253
:663.43 g/mol
:160 °C (lit.)
:13,06370
:Freezer
- Environmentally Preferable:
- Identity Test:Passes
- Purity:≥98%
- FTIR:Conforms to Standard
- pH (1% aq soln):2-4
- I max (water):259 ± 3 nm
- OD250/OD260:0.8 ± 0.15
- OD280/OD260:0.2 ± 0.15
Frequently Bought Together