Cyclizine hydrochloride 98%
Catalog # ACRO461040010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 % (120°C, 3 h)
- Titration with HClO4:≥97.5 % (on dry substance)
Specification Test Results
About this item
Synonyms: N-Benzhydryl-N'-methylpiperazine hydrochloride
:
:302.85 g/mol
Melting Pt:291…293 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS06
:GHS08
CAS Number:303-25-3
Formula:C₁₈H₂₂N₂·HCl
MDL Number:MFCD00067334