Efaproxiral sodium 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:170787-99-2
:
Formula:C₂₀H₂₂NNaO₄
MDL Number:MFCD02093864
:363.39 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Loss on drying:≤13 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together