
Gentamicin sulphate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1405-41-0
:
MDL Number:MFCD00270181
Melting Pt:218…237 °C
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- Assay:≥590 µg/mg (On dried basis)
- Loss on drying:≤18.0 % (Vacuum, 110°C, 3h)
- Sulfated ash:≤1.0 %
- pH:3.5 to 5.5 (c=4, water)
- Specific optical rotation:+107° to +121° (20°C, 589 nm) (c=1, water)
Specification Test Results
Frequently Bought Together