



Phenol 99.5% unstabilised, loose crystals, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-95-2
:
Formula:C₆H₅OH
MDL Number:MFCD00002143
:94.11 g/mol
Boiling Pt:182 °C (760 mmHg)
Melting Pt:39…42 °C
Density:1,07 g/cm³
Flash Pt:79 °C
UN:1671
ADR:6.1,II
:GHS05
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to white Loose crystals or flakes
- Infrared spectrum:Conforms
- Water:≤0.2 %
- GC:≥99.5 %
- Appearance of solution:(per Ph. Eur.) Passes test
- Residue after evaporation:≤0.05 %
- Acidity:(per Ph. Eur.) Passes test
- Solidification point:≥40.5°C
Specification Test Results
Frequently Bought Together