VWR® Water Sampling Bottles
Supplier: LAB3 OWN PRODUCTION
Certificates
About this item
Soda-Lime glass, round, clear or amber.
- Rigid and robust
- Leak proof with correct cap
- PTFE lined caps, liners fixed in so do not fall out
- Dosed with 18 mg/l sodium thiosulphate (18%)
- Expiry 12 months from manufacture
Ideal for water and general testing, filling and storage. Amber coloured for light sensitive materials.
Certifications: Standards of British ,European (2002 ) and U.S. (25) Pharmacopoeia
Amber to above and BS1679
Meet regulations 2005 (UK) and no.1935/2004 (EC) DMF No’s 10500/10501 (U.S.)articles in contact with food
Amber to above and BS1679
Meet regulations 2005 (UK) and no.1935/2004 (EC) DMF No’s 10500/10501 (U.S.)articles in contact with food
Frequently Bought Together