Chemical resistant overalls, Tychem® 6000 F Plus, model CHZ5
Supplier: DuPont
Certificates
About this item
These garments are made from a lightweight and durable fabric (<500 g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek® substrate. They offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
- Hood and self-adhesive double zipper flap
- Elasticated face, wrists, waist and ankles for an optimal fit
- Thumb loops prevent sleeves from riding up
- Self-adhesive chin flap for tight seal of suit to face-masks
- Stitched and over-taped seams with barrier tape for protection and strength
- Anti-static treatment (EN 1149-5) on inside
Tychem® 6000 F is used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.
Weight: 120 g/m²
Colour: Grey
Certifications: Type 3B/4B/5B/6B, EN 1149-5, EN 1073-2, EN 14126, Cat. III
Packaging: 25 per box, individually packed.
Specifications
- Catalog No:
- DUPOTFCHZ5TGY08/4X
- DUPOTFCHZ5TGY18/4X
- DUPOTFCHZ5TGY00/L
- DUPOTFCHZ5TGY00/M
- DUPOTFCHZ5TGY18/5X
- DUPOTFCHZ5TGY08/5X
- DUPOTFCHZ5TGY00/S
- DUPOTFCHZ5TGY26/4X
- DUPOTFCHZ5TGY26/5X
- DUPOTFCHZ5TGY00/2X
- DUPOTFCHZ5TGY18/2X
- DUPOTFCHZ5TGY26/XL
- DUPOTFCHZ5TGY18/L
- DUPOTFCHZ5TGY18/3X
- DUPOTFCHZ5TGY18/M
- DUPOTFCHZ5TGY08/3X
- DUPOTFCHZ5TGY00/5X
- DUPOTFCHZ5TGY00/3X
- DUPOTFCHZ5TGY00/XL
- DUPOTFCHZ5TGY26/3X
- DUPOTFCHZ5TGY08/S
- DUPOTFCHZ5TGY26/L
- DUPOTFCHZ5TGY08/XL
- DUPOTFCHZ5TGY08/L
- DUPOTFCHZ5TGY26/2X
- DUPOTFCHZ5TGY08/M
- DUPOTFCHZ5TGY26/M
- DUPOTFCHZ5TGY26/S
- DUPOTFCHZ5TGY08/2X
- DUPOTFCHZ5TGY18/S
- DUPOTFCHZ5TGY18/XL
- DUPOTFCHZ5TGY00/4X
- Model:
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- 6000 F Plus
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Weight:
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
Specifications
Frequently Bought Together