Organic reference standard, Trolamine, Ph.Eur., RS
Catalog # EDQMT2545000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:2.250 mg
- Standards:Reference standard
- CAS No.:102-71-6
- Description:Trolamine
- Formula:C₆H₁₅NO₃
- Storage temperature:Ambient
- Description:Trolamine
Specifications
About this item
- In Monograph 1577
- Solubility in water: 149 g/l at 20 °C
- Pharmaceutical applications