Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Triamcinolone hexacetonide, Ph.Eur., RS
Organic reference standard, Triamcinolone hexacetonide, Ph.Eur., RS
Catalog # EDQMT1950000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Triamcinolone hexacetonide, Ph.Eur., RS
Catalog # EDQMT1950000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    5611-51-8
  • Description:
    Triamcinolone hexacetonide
  • Formula:
    C₃₀H₄₁FO₇
  • Storage temperature:
    Ambient
  • Description:
    Triamcinolone hexacetonide

Specifications

About this item

  • In Monographs 0533 and 0867