Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Spiramycin, Ph.Eur., RS
Organic reference standard, Spiramycin, Ph.Eur., RS
Catalog # EDQMS1100000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Spiramycin, Ph.Eur., RS
Catalog # EDQMS1100000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    Reference standard
  • CAS No.:
    8025-81-8
  • Description:
    Spiramycin
  • Formula:
    C₄₃H₇₄N₂O₁₄
  • Storage temperature:
    Ambient
  • Description:
    Spiramycin

Specifications

About this item

  • In Monographs 0179, 0293, 1982, and 20702
  • Soluble in organic solvents
  • Partial solubility in water
  • Pharmaceutical applications