Order Entry
Ireland
ContactUsLinkComponent
Organic reference standards, Ramipril, Ph.Eur., RS
Organic reference standards, Ramipril, Ph.Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, Ramipril, Ph.Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    885516-61-0
  • Formula:
    C₂₃H₃₉ClN₂O₅
  • Storage temperature:
    Ambient
  • Description:
    Ramipril impurity C 

Specifications

Frequently Bought Together