Organic reference standards, Omeprazole, Ph.Eur., RS
Supplier: EDQM
Certificates
About this item
- In Monographs 0942, 1032, 2372, 2374, and 2787
Specifications
- CAS No.:88546-55-8
- Formula:C₁₇H₁₉N₃O₄S
- Storage temperature:Ambient
- Description:Omeprazole impurity D
Specifications
Frequently Bought Together