Order Entry
Ireland
ContactUsLinkComponent
Organic reference standards, Metoclopramide, Ph.Eur., RS
Organic reference standards, Metoclopramide, Ph.Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, Metoclopramide, Ph.Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    364-62-5
  • Formula:
    C₁₄H₂₅Cl₂N₃O₃
  • Description:
    Metoclopramide hydrochloride
  • Storage temperature:
    Ambient

Specifications

Frequently Bought Together