Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Lithocholic acid, Ph.Eur., RS
Organic reference standard, Lithocholic acid, Ph.Eur., RS
Catalog # EDQML0720800
Supplier:  EDQM
CAS Number:  
Organic reference standard, Lithocholic acid, Ph.Eur., RS
Catalog # EDQML0720800
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    60 mg
  • Standards:
    Reference standard
  • CAS No.:
    434-13-9
  • Description:
    Lithocholic acid
  • Formula:
    C₂₄H₄₀O₃
  • Storage temperature:
    Ambient
  • Description:
    Lithocholic acid

Specifications

About this item

  • In Monographs 1189 and 1275
  • Solubility in water: water: ≥ 1 g/l