Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Hyoscine butylbromide, Ph.Eur., RS
Organic reference standard, Hyoscine butylbromide, Ph.Eur., RS
Catalog # EDQMH1450000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Hyoscine butylbromide, Ph.Eur., RS
Catalog # EDQMH1450000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    20 mg
  • Standards:
    Reference standard
  • CAS No.:
    149-64-4
  • Description:
    Hyoscine butylbromide
  • Formula:
    C₂₁H₃₀BrNO₄
  • Storage temperature:
    Ambient
  • Description:
    Hyoscine butylbromide

Specifications

About this item

  • In Monograph 0737
  • Solubility in water: 33,3 - 100 g/l