Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standards, Gliclazide, Ph.Eur., RS
 :  EDQM
 
Organic reference standards, Gliclazide, Ph.Eur., RS
 :  EDQM

 

  • CAS No.:
    54786-86-6
  • Formula:
    C₇H₁₂N₂O
  • Storage temperature:
    Ambient
  • Description:
    Gliclazide impurity B

 

Frequently Bought Together