Order Entry
Ireland
ContactUsLinkComponent
Organic reference standards, Ciprofloxacin hydrochloride, Ph.Eur., RS
Organic reference standards, Ciprofloxacin hydrochloride, Ph.Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, Ciprofloxacin hydrochloride, Ph.Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    86393-33-1
  • Formula:
    C₁₃H₉ClFNO₃
  • Storage temperature:
    Ambient
  • Description:
    Ciprofloxacin impurity A

Specifications

Frequently Bought Together