Organic reference standard, Cefotaxime sodium, Ph.Eur., RS
Catalog # EDQMC0685000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:20 mg
- Standards:Reference standard
- CAS No.:64485-93-4
- Description:Cefotaxime sodium
- Formula:C₁₆H₁₆N₅NaO₇S₂
- Storage temperature:Ambient
- Description:Cefotaxime sodium
Specifications
About this item
- Pharmaceutical industry applications