Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Benzathine benzylpenicillin, Ph.Eur., RS
Organic reference standard, Benzathine benzylpenicillin, Ph.Eur., RS
Catalog # EDQMB0500000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Benzathine benzylpenicillin, Ph.Eur., RS
Catalog # EDQMB0500000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    Reference standard
  • CAS No.:
    1538-09-6
  • Description:
    Benzathine benzylpenicillin
  • Formula:
    C₄₈H₅₆N₆O₈S₂
  • Storage temperature:
    Ambient
  • Description:
    Benzathine benzylpenicillin

Specifications

About this item

Filled under inert atmosphere protected from humidity.

  • In Monograph 0373