Order Entry
Ireland
ContactUsLinkComponent
Human plasma, chronic myeloid leukemia (CML)
Human plasma, chronic myeloid leukemia (CML)
  PB006C-1-CML
 :  
Human plasma, chronic myeloid leukemia (CML)
  PB006C-1-CML
 :  
  • Pk:
    2 mL
  • Description:
    Plasma, chronic myeloid leukemia (CML), cryopreserved
Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of chronic myeloid leukemia (CML). Collected from IRB consented chronic myeloid leukemia (CML) disease donors.
No pre screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Store at −80 °C.
 : Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.