Order Entry
Ireland
ContactUsLinkComponent
Irbesartan, Reference Standards
Irbesartan, Reference Standards
Supplier:  USP
Certificates
Irbesartan, Reference Standards
Supplier:  USP

Specifications

  • CAS No.:
    748812-53-5
  • Synonym(s):
    1-pentanoylamino-cyclopentanecarboxylic acid [2'-(1H-tetrazol-5-yl)-biphenyl-4-ylmethyl]-amide
  • Formula:
    C₂₅H₃₀N₆O₂
  • Description:
    Irbesartan Related Compound A

Specifications

Frequently Bought Together