Order Entry
Ireland
ContactUsLinkComponent
Supelco® Drug and metabolite standards, prioxicam, LoGiCal®
Supelco® Drug and metabolite standards, prioxicam, LoGiCal®
Catalog # PROOLGCFOR0084.00
CAS Number:  
Supelco® Drug and metabolite standards, prioxicam, LoGiCal®
Catalog # PROOLGCFOR0084.00
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Reference standard
  • CAS No.:
    36322-90-4
  • Description:
    Piroxicam
  • Formula:
    C₁₅H₁₃N₃O₄S
  • Description:
    Piroxicam

Specifications

About this item