Human recombinant sterol O-acyltransferase 2 (from E. coli)
Catalog # PRSI92-209
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Environmentally Preferable:
- Tag sequence:N-Trx, 6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Sterol O-acyltransferase 2
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:59.1 kD
- Sequence:Met1-Glu397
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM PB,150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Biological assays
Specifications
About this item
ACAT2 is a cytoplasmic enzyme which belongs to the thiolase family. ACAT2 takes part in lipid metabolism, lipoprotein assembly, catalyzing cholesterol esterification in mammalian cells. It is responsible for the synthesis of cholesteryl esters which are part of lipoproteins containing apoB. ACAT2 deficiency contributes to severe mental retardation and hypotonus.
Fusion-Tag: N-Trx, 6 His tag
This recombinant protein can be used for biological assays. For research use only.