Mouse recombinant periostin (from CHO cells)
PRSI90-267
:
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Mouse
- Tag sequence:DDDDK Tag
- Storage conditions:Stable for at least 6 months after receipt when stored at −20 °C.
- Protein synonyms:OSF-2|Postn|Osteoblast-specific Factor 2
- Protein/peptide name:Periostin
- Purity:>95% (SDS-PAGE)
- Sequence:24-783
- Formulation:Lyophilized, Contains PBS.
Periostin was originally isolated as an osteoblast-specific factor that functions as a cell adhesion molecule for pre-osteoblasts and is thought to be involved in osteoblast recruitment, attachment and spreading. Additionally, periostin expression has previously been shown to be significantly increased by both transforming growth factor beta1 (TGF-beta1) and bone morphogenetic protein 2 (BMP-2). Abnormal expression of Periostin is also linked to angiogenesis and metastasis in epithelial tumours.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.