- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:SERPINB12|Serpin B12
- Protein/peptide name:Serpin B12
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:49.5 kD
- Sequence:Met1-Pro425
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Serpin B12 is a member of the serpin family. Serpins are the largest and most diverse family of serine protease inhibitors. Most serpins are secreted and attain physiologic concentrations in the blood and extracellular fluids. Serpin B12 is expressed in many tissues, including brain, bone marrow, lymph node, heart, lung, liver, pancreas, testis, ovary, and intestine. Serpins are involved in a number of fundamental biological processes such as blood coagulation, complement activation, fibrinolysis, angiogenesis, inflammation and tumour suppression and are expressed in a cell-specific manner. SerpinB12 inhibits trypsin and plasmin, but not thrombin, coagulation factor Xa, or urokinase-type plasminogen activator.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.