Human recombinant UBE2D1 (from E. coli)
Catalog # PRSI92-272
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-GST tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Ubiquitin-conjugating enzyme E2(17)KB 1|Ubiquitin-protein ligase D1|UBCH5A|UBC4/5 homolog|Ubiquitin carrier protein D1|UbcH5|SFT|UBC5A|Ubiquitin-conjugating enzyme E2 D1|Stimulator of Fe transport|Ubiquitin-conjugating enzyme E2-17 kDa 1
- Protein/peptide name:UBE2D1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:42.9 kD
- Sequence:Met1-Met147
- Formulation:Lyophilized from a 0.2 um filtered solution of 50mM HEPES,150mM NaCl,2mM DTT,10% Glycerin pH7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Ubiquitin-conjugating enzyme E2 D1(UBE2D1)belongs to the ubiquitin-conjugating enzyme family. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This enzyme is closely related to a stimulator of iron transport (SFT), and is up-regulated in hereditary hemochromatosis. It also functions in the ubiquitination of the tumor-suppressor protein p53 and the hypoxia-inducible transcription factor HIF1alpha by interacting with the E1 ubiquitin-activating enzyme and the E3 ubiquitin-protein ligases.
Fusion-Tag: N-GST tag
This recombinant protein can be used for biological assays. For research use only.