Order Entry
Ireland
ContactUsLinkComponent
 
Human recombinant MGAT2 (from cells)
  PRSI91-651
 :  
Human recombinant MGAT2 (from cells)
  PRSI91-651
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    6-Mannosyl-Glycoprotein 2-Beta-N-Acetylglucosaminyltransferase|Alpha-1|Beta-1|MGAT2|Mannoside Acetylglucosaminyltransferase 2|GlcNAc-T II|N-Glycosyl-Oligosaccharide-Glycoprotein N-Acetylglucosaminyltransferase II|2-N-acetylglucosaminyltransferase II|NT-II|GlcNAc-TII
  • Protein/peptide name:
    MGAT2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    49.3 kD
  • Sequence:
    Arg30-Gln447
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl,150mM NaCl, pH8.0 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Mannoside Acetylglucosaminyltransferase 2 (MGAT2) is a single-pass type II membrane protein that contains the typical glycosyltransferase domains: a short N-terminal cytoplasmic domain, a hydrophobic non-cleavable signal-anchor domain and a C-terminal catalytic domain. MGAT2 catalyzes an essential step in the conversion of oligo-mannose to complex N-glycans. Defects in MGAT2 are the cause of congenital disorder of glycosylation type 2A.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag