Order Entry
Ireland
ContactUsLinkComponent
Human recombinant VEGF165 (from HEK293 cells)
Human recombinant VEGF165 (from HEK293 cells)
Catalog # PRSI96-773
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant VEGF165 (from HEK293 cells)
Catalog # PRSI96-773
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Environmentally Preferable:
  • Gene ID:
    7422
  • Protein synonyms:
    RP1-261G23.1|MVCD1|MGC70609|VEGFA|VEGF165|VPF
  • Protein/peptide name:
    VEGF165
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    20 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

Specifications

About this item

VEGF165 is the most abundant splice variant of VEGF-A. VEGF165 is produced by a number of cells including endothelial cells, macrophages and T cells. VEGF165 is involved in angiogenesis, vascular endothelial cell survival, growth, migration and vascular permeability. VEGF gene expression is induced by hypoxia, inflammatory cytokines and oncogenes. VEGF165 binds to heparan sulfate and is retained on the cell surface and in the extracellular matrix. VEGF165 binds to the receptor tyrosine kinases, VEGFR1 and VEGFR2. VEGF165 is the only splice variant that binds to co-receptors NRP-1 and NRP-2 that function to enhance VEGFR2 signaling. Binding of VEGF165 to VEGFR1 and VEGFR2 leads to activation of the PI3K/AKT, p38 MAPK, FAK and paxillin. VEGF plays a key role in tumor angiogenesis in many cancers.