Order Entry
Ireland
ContactUsLinkComponent
Human Recombinant CD2001 (from HEK293 cells)
Human Recombinant CD2001 (from HEK293 cells)
Catalog # PRSI96-573
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human Recombinant CD2001 (from HEK293 cells)
Catalog # PRSI96-573
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilized recombinant human CD200R1 at 2μg/ml,the concentration of rhCD200 that produces 50% of the optimal binding response is appoximately 0. 5‑16 ng/mL.
  • Protein synonyms:
    OX-2|MOX1|My033|MRC|MOX2|CD200
  • Protein/peptide name:
    CD2001
  • Purity:
    >98% as determined by SDS-PAGE.
  • Molecular weight:
    23.4 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

Specifications

About this item

CD200 is also known as OX-2 membrane glycoprotein (OX-2), is a type-1 membrane glycoprotein, which contains two immunoglobulin domains (1 Ig-like C2-type (immunoglobulin-like) domain and Ig-like V-type (immunoglobulin-like) domain), and thus belongs to the immunoglobulin superfamily. CD200 / OX-2 is widely expressed in multiple cell types. CD200 interacts with a structurally related receptor (CD200R) expressed mainly on myeloid cells and is involved in regulation of macrophage and mast cell function. OX-2 / CD200 and CD200R associate via their respective N-terminal Ig-like domains. CD200 also plays an important role in prevention of graft rejection, autoimmune diseases and spontaneous abortion.