Human recombinant MICA (from HEK293 cells)
Catalog # PRSI96-536
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Environmentally Preferable:
- Protein synonyms:MIC-A
- Protein/peptide name:MICA
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:33.6 kDa
- Formulation:Lyophilized, PBS, pH7.4
- Tested applications:Western Blot
Specifications
About this item
MHC class I polypeptide-related sequence A (MICA) belongs to the MHC class I family and MIC subfamily. MICA contains one Ig-like C1-type (immunoglobulin-like) domain. Unlike classical MHC class I molecules, MICA does not form a heterodimer with beta-2-microglobulin. MICA acts as a stress-induced self-antigen that is recognized by gamma delta T-cells. MICA is ligand for the KLRK1/NKG2D receptor. MICA bind to KLRK1 leads to cell lysis.