Order Entry
Ireland
ContactUsLinkComponent
Human recombinant Fetuin A (from HEK293 cells)
Human recombinant Fetuin A (from HEK293 cells)
  PRSI96-311
 :  
undefined
Human recombinant Fetuin A (from HEK293 cells)
  PRSI96-311
 :  

 

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Protein synonyms:
    Fetuin A|FETUA|AHSG
  • Protein/peptide name:
    Fetuin A
  • Purity:
    >98% as determined by SDS-PAGE.
  • Molecular weight:
    38.2 kDa
  • Formulation:
    Lyophilized, 25 mM Tris and 150 mM NaCl, pH 7.5
  • Tested applications:
    Western Blot

 

 

Alpha-2-HS-glycoprotein (AHSG) is also known as Alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, Fetuin-A (FETUA), is a glycoprotein that is composed of two subunits, the A and B chains, belongs to the cystatin family of proteases inhibitors and the fetuin class of plasma binding proteins. AHSG / Fetuin A is highly expressed in embryonic cells and adult hepatocytes, and is expressed to a lesser extent in monocytes/macrophages. Fetuins are carrier proteins like albumin. Fetuin-A / AHSG forms soluble complexes with calcium and phosphate and thus is a carrier of otherwise insoluble calcium phosphate. Thus AHSG / Fetuin A is a potent inhibitor of pathological calcification. Fetuin A / FETUA promotes phagocytosis of apoptotic cells and plays a role in bone development, calcium homeostasis and insulin sensitivity. Mice deficient in Fetuin-A show systemic calcification of soft tissues.