Anti-TNF Mouse Monoclonal Antibody [clone: 2.1_4E10-1H11]
Catalog # PRSIXP-5711-M
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Tumor Necrosis Factor Alpha
- Antigen symbol:TNFA
- Clonality:Monoclonal
- Clone:2.1_4E10-1H11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Neutralisation:Yes
- Form:Lyophilized
- Gene ID:7124
- Antigen synonyms:tumor necrosis factor ligand superfamily member 2|tumor necrosis factor alpha|DIF|monocyte-derived|Tnfa|cachectin|Tnfsf1a|TNFalpha|member 2|TNF-alpha|TNF-a|TNFSF2|macrophage-derived|TNF superfamily|TNF|tumor necrosis factor-alpha|APC1 protein
- Storage temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping temperature:RT
- Immunogen:The monoclonal antibody was produced in BALB/c X ICR F1 mice using recombinant hTNF-alpha as the immunizing antigen.
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Tumor Necrosis Factor Alpha (TNF alpha) is a protein secreted by lipopolysaccharide stimulated macrophages, and causes tumor necrosis when injected into tumour bearing mice. TNF alpha is believed to mediate pathogenic shock and tissue injury associated with endotoxemia.
Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: 2.1_4E10-1H11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human