- Antibody type:Primary
- Antigen name:inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta
- Antigen symbol:IKBKB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:3551
- Antigen synonyms:I-kappa-B kinase 2|IKK-B|IkBKB|I-kappa-B-kinase beta|IKK2
- Modification:phospho-Tyr188
- Storage buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Molecular weight:87 kDa
- Storage temperature:Store antibody at –20 °C for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:-20°C
- Immunogen:IKK- beta (phospho Tyr188) antibody was raised against a peptide sequence around phosphorylation site of tyrosine 188 (L-Q-Y (p)-L-A) derived from Human IKK- beta.
- Purification:Purified by affinity-chromatography using epitope-specific peptide.
- Size:100 µg
- Pk:100 µG
Serine kinase that plays an essential role in the NF-kappa-B signaling pathway which is activated by multiple stimuli such as inflammatory cytokines, bacterial or viral products, DNA damages or other cellular stresses. Acts as part of the canonical IKK complex in the conventional pathway of NF-kappa-B activation and phosphorylates inhibitors of NF-kappa-B on 2 critical serine residues. These modifications allow polyubiquitination of the inhibitors and subsequent degradation by the proteasome. In turn, free NF-kappa-B is translocated into the nucleus and activates the transcription of hundreds of genes involved in immune response, growth control, or protection against apoptosis. In addition to the NF-kappa-B inhibitors, phosphorylates several other components of the signaling pathway including NEMO/IKBKG, NF-kappa-B subunits RELA and NFKB1, as well as IKK-related kinases TBK1 and IKBKE. IKK-related kinase phosphorylations may prevent the overproduction of inflammatory mediators since they exert a negative regulation on canonical IKKs. Also phosphorylates other substrates including NCOA3, BCL10 and IRS1. Within the nucleus, acts as an adapter protein for NFKBIA degradation in UV-induced NF-kappa-B activation.
Western Blot: 1:500~1:1000
Type: Primary Antibody
Antigen: IKBKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG
Reactivity: Human, Mouse, Rat