- Antigen symbol:CCL17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Concentration:1 mg/mL
- Immunogen:Rabbit polyclonal CCL17 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human CCL17.
- Size:100 µg
- Pk:100 µG
CCL17 Antibody: CCL17, also known as TARC, binds to chemokine receptors CCR4 and CCR8 and plays important roles in T cell development in thymus as well as in trafficking and activation of mature T cells. CCL17 is secreted from monocyte-derived DCs and endothelial cells and is responsible for selective recruitment and migration of activated Th2 lymphocytes. CCL17 is also required for CCR7 and CXCR4-dependent migration of cutaneous dendritic cells under inflammatory conditions.
CCL17 Antibody is affinity chromatography purified via peptide column.
CCL17 Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CCL17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat