Anti-XIAP Rabbit Polyclonal Antibody
Catalog # PRSI70-833
Supplier: ProSci Inc.
Specifications
- Antigen name:X-linked inhibitor of apoptosis
- Antigen symbol:XIAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:IAP-like protein|HILP|API3|Baculoviral IAP repeat-containing protein 4|X-linked IAP|X-linked inhibitor of apoptosis protein|XIAP|IAP3|Inhibitor of apoptosis protein 3
- Concentration:1 mg/mL
- Immunogen:Recombinant protein fragment containing a sequence corresponding to a region within amino acids 75 and 226 of BIRC4.
- Tested applications:Immunoaffinity Chromatography
- Size:50 µl
- Pk:50 µl
Specifications
About this item
XIAP Antibody can be used for Immunocytochemistry starting at 1:100 - 1:200, Immunofluorescence starting at 1:100 - 1:200, Immunohistochemistry in paraffin embeded tissues starting at 10μg/mL, and Western Blotting starting at 1:500 - 1:1000.
0.1 M Tris-glycine, pH 7, 10% Glycerol, 0.01% Thimerosal
0.1 M Tris-glycine, pH 7, 10% Glycerol, 0.01% Thimerosal
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: XIAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human