Anti-ABCD1 Rabbit Polyclonal Antibody
Catalog # PRSI70-396
Supplier: ProSci Inc.
Specifications
- Antigen name:ATP-binding cassette, Subfamily D (ALD), Member 1
- Antigen symbol:ABCD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Antigen synonyms:ALD|ABC42|ALDP|AMN
- Concentration:1 mg/mL
- Immunogen:Recombinant protein
- Size:50 µl
- Pk:50 µl
Specifications
About this item
ABCD1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 μg/mL, and Western Blotting.
Affinity Purified
PBS, pH 7.4, 0.02% sodium azide
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ABCD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse