Anti-IL17A Rat Monoclonal Antibody
Catalog # PRSI51-842
Supplier: ProSci Inc.
Specifications
- Antigen name:Interleukin 17 A
- Antigen symbol:IL17A
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:IL-17|IL17|CTLA8|IL-17A|CTLA-8|Cytotoxic T-lymphocyte-associated serine esterase 8
- Tested applications:Fluorescence-Activated Cell Sorting
- Size:100 µg
- Pk:100 µG
Specifications
About this item
IL-17 antibody can be used in Western Blot, and immunohistochemistry starting at 1:50.
Purified
Borate buffered saline.
Aliquot and store IL-17 antibody at -20 °C. As with all antibodies avoid freeze/thaw cycles.
Type:
Antigen: IL17A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse