Specifications
- Antigen name:P21 Protein (Cdc42/Rac)-Activated Kinase 1
- Antigen symbol:PAK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:p65-PAK|MGC130000|Alpha-PAK|MGC130001|PAK-1|PAK1|p21/Cdc42/Rac1-activated kinase 1. PAK alpha
- Immunogen:Phosphopeptide corresponding to amino acid residues surrounding the phospho-Thr402 of rat p21 activated kinase 2 (PAK-2). The peptide sequence used is identical in PAK-1, 2 and 3. Note: Thr402 in PAK-2 corresponds to Thr423 in human PAK-1.
- Size:100 µl
- Pk:100 µl
Specifications
About this item
In mammals, there are several identified isoforms of p21-activated protein kinases or PAKs: alpha-PAK (also known as PAK-1) and beta-PAK (also known as PAK-3) are mostly brain-specific, while -PAK (also known as PAK-2) is expressed ubiquitously (Jakobi et al., 2003). Mutations of the gene coding for PAK-3 are associated with X-linked mental retardation and recent work indicates that PAK-3 is a key regulator of synapse formation and plasticity in the hippocampus (Boda et al., 2004). PAK-3 is thought to play a key role in regulation of cell shape and motility as well as cell death (Jakobi et al., 2003; Walter et al., 1998). Autophosphorylation of Thr402 in the protein has been found to be essential for activation of PAK (Jakobi et al., 2000).
The antibody has been directly tested for reactivity in Western blots with rat tissue. It is anticipated that the antibody will react with bovine, canine, human and mouse based on the fact that these species have 100% homology with the amino acid sequence used as antigen.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Bovine