Order Entry
Ireland
ContactUsLinkComponent
 
Anti-GRIN2B Rabbit Polyclonal Antibody
Catalog # PRSI50-232
Supplier:  ProSci Inc.
undefined
Anti-GRIN2B Rabbit Polyclonal Antibody
Catalog # PRSI50-232
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Glutamate Ionotropic Receptor NMDA Type Subunit 2B
  • Antigen symbol:
    GRIN2B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Zebrafish,
    Chicken,
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    hNR3|GluN2B|NR2B|EIEE27|MRD6|NMDAR2B|glutamate ionotropic receptor NMDA type subunit 2B
  • Immunogen:
    Phosphopeptide corresponding to amino acid residues surrounding the phospho-Tyr1252 of the NR2B subunit of the rat NMDA receptor.
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

The NMDA receptor (NMDAR) plays an essential role in memory, neuronal development and it has also been implicated in several disorders of the central nervous system including Alzheimer’s, epilepsy and ischemic neuronal cell death (Grosshans et al., 2002; Wenthold et al., 2003; Carroll and Zukin, 2002). The rat NMDAR1 (NR1) was the first subunit of the NMDAR to be cloned. The NR1 protein can form NMDA activated channels when expressed in Xenopus oocytes but the currents in such channels are much smaller than those seen in situ. Channels with more physiological characteristics are produced when the NR1 subunit is combined with one or more of the NMDAR2 (NR2 A-D) subunits (Ishii et al., 1993). Phosphorylation of Tyr1252 is thought to potentiate NMDA receptor-dependent influx of calcium (Takasu et al., 2002).

The antibody has been directly tested for reactivity in Western blots with rat tissue. It is anticipated that the antibody will react with bovine, canine, chicken, human, mouse, non-human primate and zebra fish based on the fact that these species have 100% homology with the amino acid sequence used as antigen.

Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Grin2b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Zebrafish, Bovine, Chicken