Specifications
- Antigen name:Gamma-Aminobutyric Acid (GABA) B Receptor, 2
- Antigen symbol:GABBR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Xenopus,Chicken,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:GABABR2|HRIHFB2099|GPRC3B|GPR51|HG20
- Immunogen:Phosphopeptide corresponding to amino acid residues surrounding the phospho-Ser783 of GABAB R2.
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Gamma-aminobutyric acid (GABA) is the primary inhibitory neurotransmitter in the central nervous system. There are two major classes of GABA receptors: the GABAA and the GABAB subtype of receptors. GABAB receptors are heterodimeric G protein-coupled receptors that mediate slow synaptic inhibition in the central nervous system. It has recently been demonstrated that AMPK binds directly to GABAB receptors and phosphorylates S783 in the cytoplasmic tail of the R2 subunit and that S783 plays a critical role in enhancing neuronal survival after ischemia as phosphorylation of S783 is evident in many brain regions and is increased dramatically after ischemic injury to the brain (Kuramoto et al., 2007).
The antibody has been directly tested for reactivity in Western blots with rat tissue. It is anticipated that the antibody will react with mouse, human, non-human primate, chicken, bovine and Xenopus tissues based on the fact that these species have 100% homology with the amino acid sequence used as antigen.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Gabbr2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Bovine, Chicken, Xenopus