- Antigen name:Glutamate Receptor 2
- Antigen symbol:GRIA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Zebrafish,Chicken,Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:MGR3|GLURB|glutamate receptor|GluR-2|GRM3|AMPA-selective glutamate receptor 2|Glutamate receptor ionotropic|GluR-K2|ionotropic|GRIA2|glutamate receptor metabotropic type 3|HBGR2|Glutamate receptor 2|GPRC1C|GLUR2|AMPA 2|GluR-B|Metabotropic glutamate receptor 3 precursor
- Immunogen:Peptide corresponding to amino acid residues from rat GluR2, conjugated to keyhole limpet hemocyanin (KLH).
- Size:100 µl
- Pk:100 µl
The ion channels activated by glutamate are typically divided into two classes. Those that are sensitive to N-methyl-D-aspartate (NMDA) are designated NMDA receptors (NMDAR) while those activated by alpha-amino-3-hydroxy-5-methyl-4-isoxalone propionic acid (AMPA) are known as AMPA receptors (AMPAR). The AMPAR are comprised of four distinct Glutamate Receptor Subunits designated (GluR1-4) and they play key roles in virtually all excitatory neurotransmission in the brain (Keinänen et al., 1990;Hollmann and Heinemann, 1994). The GluR2 subunit is thought to play a key role in forms of synaptic plasticity such as LTD (Chung et al., 2003)
The antibody has been directly tested for reactivity in Western blots with human tissue. It is anticipated that the antibody will also react with chicken, human, mouse, non-human primates and zebra fish based on the fact that these species have 100% homology with the amino acid sequence used as antigen.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Gria2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Chicken, Zebrafish